Learn More
Strem, An Ascensus Company CAS# 13477-34-4. 1kg. Calcium nitrate tetrahydrate, 99%. MFCD00149604
SDP
Catalog No. 5090114368
Encompass
CAS# 13477-34-4. 1kg. Calcium nitrate tetrahydrate, 99%. MFCD00149604. Molecular Weight: 164.09 (236.16). Molecular Formula: Ca(NO3)2∙4H2O. Color/form: white xtl. Strem# 93-2028. http://www.strem.com/catalog/v/93-2028/
Catalog No. 50-901-14368 Supplier Strem, An Ascensus Company Supplier No. 9320281000G
CAS# 13477-34-4. 1kg. Calcium nitrate tetrahydrate, 99%. MFCD00149604. Molecular Weight: 164.09 (236.16). Molecular Formula: Ca(NO3)2∙4H2O. Color/form: white xtl. Strem# 93-2028. http://www.strem.com/catalog/v/93-2028/