Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Calcium nitride

Catalog No. 7564155G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-641-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75-641-55G Supplier Sigma Aldrich Supplier No. 7564155G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Calcium nitride is used as a nitride source in metathesis reactions to produce complex nitrides as well as SiAlON-related optical materials.

Specifications

CAS 12013-82-0
Density 2.63 g/mL (at 25°C)
Molecular Formula Ca3N2
Linear Formula Ca3N2
Quantity 5 g
MDL Number MFCD00015985
Synonym Tricalcium dinitride
Molecular Weight (g/mol) 148.25
Percent Purity 99% trace metals basis (contains <0.5%Mg)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.