Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium nitride, 99% (metals basis)

Catalog No. AA1312430
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1312430 250 g
AA1312406 5 g
AA1312418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1312430 Supplier Thermo Scientific Chemicals Supplier No. 01312430
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12013-82-0 | Ca3N2

Calcium nitride is used in the production of light-emitting diode (LED) phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12013-82-0
Molecular Formula Ca3N2
MDL Number MFCD00015985

Specifications

Melting Point 1195°C
Color Brown
Physical Form Pieces
Quantity 250 g
Assay Percent Range 99% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 148.25
Odor Ammonia-like
Sensitivity Moisture sensitive
Density 2.63
Chemical Name or Material Calcium nitride
Show More Show Less
Hazard Category H228-H314-H335
Precautionary Statement P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation DOT Class: 4.1, Packaging Group: II
EINECSNumber 234-592-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.