Learn More
Eurofins Discoverx Calcium No WashPLUS, 10 plates (384-well). (90-0091)
Catalog No. 501999540
Encompass
The Calcium No WashPLUS detection kit is intended for use with cells expressing receptors that signal through calcium. The kit contains a calcium-sensitive dye which assays intracellular calcium levels upon activation of a Gαq-coupled GPCR. Activation of the GPCR leads to release of intracellular calcium, and a detectable increase in fluorescent signal. This detection kit contains reagents for assaying 10 plates. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/cell-line/calcium-no-washplus-90-0091.
Catalog No. 50-199-9540 Supplier Eurofins Discoverx Supplier No. 900091
The Calcium No WashPLUS detection kit is intended for use with cells expressing receptors that signal through calcium. The kit contains a calcium-sensitive dye which assays intracellular calcium levels upon activation of a Gαq-coupled GPCR. Activation of the GPCR leads to release of intracellular calcium, and a detectable increase in fluorescent signal. This detection kit contains reagents for assaying 10 plates. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/cell-line/calcium-no-washplus-90-0091.