Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium Oxide
SCI_ED

Catalog No. S25227 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade - Powder
Catalog No. Quantity Packaging
S25227 100 g Lab Grade - Powder
S25227A 500 g Lab Grade - Powder
2 options

Catalog No. S25227

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
PubChem CID 14778
ChEBI CHEBI:31344
IUPAC Name calcium oxidandiide
SMILES [O--].[Ca++]

Specifications

Color Beige-Yellow to White
Quantity 100 g
Assay Percent Range >96% w/v
Linear Formula CaO
Formula Weight 56.08
Grade Laboratory
Packaging Lab Grade - Powder
Density 3.33g/mL
Chemical Name or Material Calcium Oxide
ShelfLife 12 months
DOTInformation DOT Class 8, III, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.