Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Calcium oxide

Catalog No. 20815925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-815-925G 25 g
20-815-9500G 500 g
20-815-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 20-815-925G Supplier Sigma Aldrich Supplier No. 20815925G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Units are 10% in excess of stated weight to compensate for the loss of H2O and CO2ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 1305-78-8
Density 3.3 g/mL (at 25°C)
Boiling Point 2850°C
Molecular Formula CaO
Linear Formula CaO
Quantity 25 g
MDL Number MFCD00010911
Synonym Lime; Quicklime
Molecular Weight (g/mol) 56.08
Percent Purity 99.9% trace metals basis
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.