Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Oxide, Solstice

Catalog No. 60044713 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
6 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-713 6 x 1 kg
60-019-19 1 kg
60-019-21 5 kg
60-019-20 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-044-713 Supplier Solstice Supplier No. 120476X1KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss., meets analytical specification of FCC, 96-100.5% (ex ignited substance), powder (fine)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
PubChem CID 14778
ChEBI CHEBI:31344
SMILES [O--].[Ca++]

Specifications

Quantity 6 x 1 kg
Density 3.3g/mL at 25°C
Linear Formula CaO
UN Number UN1910
IUPAC Name calcium oxidandiide
Formula Weight 56.08
Percent Purity 96 to 100.5% (ex ignited substance)
Grade puriss.
Chemical Name or Material Calcium Oxide, Meets analytical specification of FCC
Hazard Statement P261-P280-P305 + P351 + P338
Precautionary Statement H315-H318-H335
DOTInformation PG 8 - Calcium oxide
EINECSNumber 215-138-9
RTECSNumber EW3100000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.