Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Oxide, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

1305-78-8
CaO
56.077
ODINCKMPIJJUCX-UHFFFAOYSA-N
14778
CHEBI:31344

High quality chemicals for laboratory and specialized industrial use

CAS 1305-78-8
Molecular Formula CaO
InChI Key ODINCKMPIJJUCX-UHFFFAOYSA-N
SMILES O=[Ca]
IUPAC Name oxocalcium
Molecular Weight (g/mol) 56.077
PubChem CID 14778
ChEBI CHEBI:31344
Formula Weight 56.08
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 3.37kg
Quantity 500g
Chemical Name or Material Calcium Oxide
Grade BAKER ANALYZED™ Reagent
Show More Show Less