Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Oxide, Powder, Technical, Spectrum™ Chemical
SDP

Catalog No. 18601331 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-331 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-601-331 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C11412.5KG

missing translation for 'validMainframeMsg'

C1141, 1305-78-8, CaO

Spectrum™ Chemical Calcium Oxide, Powder, Technical , is a widely used chemical compound known as quicklime. A white, alkaline crystalline solid that is also caustic. It is a component of common cement and used in the fabrication of paper and plaster. In the petroleum industry, calcium oxide and phenolpthalein are combined into a water detection paste .

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
IUPAC Name calcium oxidandiide
SMILES [O--].[Ca++]

Specifications

CAS Max % 100%
Quantity 2.5 kg
UN Number UN1910
Grade Technical
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.