Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Oxide, Purified (Lime), Reagents

Catalog No. 01337330
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-330 3 kg
01-337-329 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-330 Supplier Reagents Holdings Llc Supplier No. C21660003KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calcium Oxide, Purified (Lime), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym Lime, Quicklime, Burnt lime
IUPAC Name calcium oxidandiide
SMILES [O--].[Ca++]

Specifications

Boiling Point 2850°C
Melting Point 2570°C
Color White to Off-White
Physical Form Solid
Quantity 3 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 3.34
Chemical Name or Material Calcium Oxide, Purified (Lime)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.