Learn More
U.S. Pharmacopeia Calcium Pantothenate, 137-08-6, MFCD00002766, 300mg
Catalog No. NC0088410
Encompass
Molecular formula C18H32CaN2O10, Molecular weight 476.53, Melting Point 383 - 384.8 °F (195 - 196 °C) (decomposes), Vitamin B5, Pantothenic acid calcium salt
Catalog No. NC0088410 Supplier U.S. Pharmacopeia Supplier No. 1087009
Molecular formula C18H32CaN2O10, Molecular weight 476.53, Melting Point 383 - 384.8 °F (195 - 196 °C) (decomposes), Vitamin B5, Pantothenic acid calcium salt