Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Pantothenate, Powder, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601336 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-336 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-601-336 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CA159100GM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

CA159, 137-08-6, C18H32CaN2O10

Spectrum™ Chemical Calcium Pantothenate, Powder, USP is often used in dietary supplements a more stable of pantothenic acid (Vitamin B5) in the digestive tract. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 137-08-6
Molecular Formula C18H32CaN2O10
Molecular Weight (g/mol) 476.54
MDL Number MFCD00002766
InChI Key FAPWYRCQGJNNSJ-DXHDTSSINA-L
IUPAC Name calcium bis(3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanoate)
SMILES [Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O

Specifications

CAS Max % 100%
Quantity 100 g
Percent Purity 98 to 102%
Grade USP
Loss on Drying 5%
Optical Rotation +25.0° to +27.5°
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.