Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Peroxide, FCC, 60%, Spectrum™ Chemical
SDP

Catalog No. 18601344 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
100 lb.
Packaging:
Amber Glass Bottle
Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-344 12 kg Poly Pail
18-601-341 25 g Amber Glass Bottle
18-601-342 125 g Amber Glass Bottle
18-601-343 500 g Amber Glass Bottle
18-601-345 100 lb. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-344 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C138812KG
Only null left
Add to Cart
Add to Cart

C1388, 1305-79-9, CaO2

Spectrum™ Chemical Calcium Peroxide, FCC is used as a food additive under the E number E930 as flour bleaching agent. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1305-79-9
Molecular Formula CaO2
Molecular Weight (g/mol) 72.08
InChI Key LHJQIRIGXXHNLA-UHFFFAOYSA-N
IUPAC Name calcium dioxidanediide
SMILES [Ca++].[O-][O-]

Specifications

CAS Max % 100%
UN Number UN1457
Quantity 12 kg
Percent Purity 60%
Grade FCC
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.