Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium phosphate, dibasic, 97%, pure, anhydrous

Catalog No. AC215710010
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215710010 1 kg Plastic bottle
AC215710250 25 g Plastic bottle
AC215710050 5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC215710010 Supplier Thermo Scientific Chemicals Supplier No. 215710010
Only null left
Add to Cart

CAS: 7757-93-9 | CaHO4P | 136.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
Synonym calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
PubChem CID 24441
ChEBI CHEBI:32596
IUPAC Name calcium;hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 96% min. (Complexometry)
Linear Formula CaHPO4
Merck Index 15,1694
Solubility Information Solubility in water: 0.1g/L (25°C). Other solubilities: practically insoluble in alcohol
Formula Weight 136.06
Percent Purity 97%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Calcium phosphate, dibasic
Show More Show Less
EINECSNumber 231-826-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.