Learn More
Alkali Scientific Calcium Phosphate, Dibasic, Dihydrate, 500 G
SDP
Catalog No. 5000660858
Encompass_Preferred
Calcium Phosphate, Dibasic, Dihydrate is a laboratory-grade reagent available in a 500g quantity, widely used in biochemical applications. This compound is commonly used to prepare buffer solutions and in molecular biology procedures, particularly in cell culture media and DNA research. Calcium phosphate plays a pivotal role in cellular activities such as bone formation, cellular signaling, and protein synthesis. It is often employed in transfection protocols, where it helps facilitate the uptake of DNA into mammalian cells. As a source of both calcium and phosphate, it is essential in studies of mineralization and metabolism within biological systems.
Catalog No. 50-006-60858 Supplier Alkali Scientific Supplier No. ATE002CP
Calcium Phosphate, Dibasic, Dihydrate is a laboratory-grade reagent available in a 500g quantity, widely used in biochemical applications. This compound is commonly used to prepare buffer solutions and in molecular biology procedures, particularly in cell culture media and DNA research. Calcium phosphate plays a pivotal role in cellular activities such as bone formation, cellular signaling, and protein synthesis. It is often employed in transfection protocols, where it helps facilitate the uptake of DNA into mammalian cells. As a source of both calcium and phosphate, it is essential in studies of mineralization and metabolism within biological systems.