Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium Phosphate Transfec
Catalog No. 502906132
Encompass
Calcium phosphate transfection is a commonly used method for the introduction of DNA into eukaryotic cells. This technique has been used to obtain both transient and stable transfections in a wide variety of cell types.
Catalog No. 50-290-6132 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CAPHOS1KT
Calcium phosphate transfection is a commonly used method for the introduction of DNA into eukaryotic cells. This technique has been used to obtain both transient and stable transfections in a wide variety of cell types.