Learn More
Bioworld Calcium Phosphate, Tribasic, 250 g
Catalog No. 50255701
Encompass_Preferred
Synonym: Calcium Hydroxyphosphate, HAp, HydroxyapatiteFor preparing hydroxyapatite columns, please use catalog number # 40800054. Do not use this catalog number for preparing columns.Grade: ReagentAppearance: PowderMelting Point: 1100 °C(lit.)
Catalog No. 50-255-701 Supplier Bioworld Supplier No. 403001532
Synonym: Calcium Hydroxyphosphate, HAp, HydroxyapatiteFor preparing hydroxyapatite columns, please use catalog number # 40800054. Do not use this catalog number for preparing columns.Grade: ReagentAppearance: PowderMelting Point: 1100 °C(lit.)