Learn More
Calcium phosphate tribasic, 34-40% Ca
Description
Utilized in powdered spices as an anticaking agent and baby powder. It is the starting material for the production of phosphoric acid and fertilizers. Employed in tissue replacement for repairing bony defects and act as drug carrier for drug delivery to the bone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 10 kg |
| Assay Percent Range | 34-40% Ca |
| Solubility Information | Partially soluble in water. |
| Formula Weight | 1004.67 |
| Appearance | White powder |
| Chemical Name or Material | Calcium phosphate tribasic, tribasic |
Safety and Handling
| Hazard Category | H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| EINECSNumber | 235-330-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.