Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phosphate Tribasic (Powder/FCC/NF), Fisher Chemical

Catalog No. C133500 Shop All Fisherbrand Products
Change view
Click to view available options
Poly Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
C133-500 Poly Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C133-500

missing translation for 'validMainframeMsg'

Specifications

Melting Point 1670°C
Color White
CAS 7758-87-4
Physical Form Solid
Quantity 500 g
Molecular Formula Ca3O8P2
MDL Number MFCD00010904
Synonym calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]
IUPAC Name tricalcium;diphosphate
Molecular Weight (g/mol) 310.174
PubChem CID 24456
ChEBI CHEBI:9679
Formula Weight 502.31
Percent Purity 34.0 to 40.0% (FCC)
Grade FCC/NF
Identification Pass Test (FCC), Pass Test (FCC/NF), Pass Test (NF)
Loss on Ignition 10.0% max. (FCC), 10.0% max. (FCC), 8.0% max. (NF), 8.0% max. (NF)
Packaging Poly Bottle
Density 3.14g/cm³
Chemical Name or Material Calcium Phosphate Tribasic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.