Learn More
Medchemexpress LLC Calcium phosphorylcholine chloride | 4826-71-5 | 98.95% | 25 G
Supplier: Medchemexpress LLC 25GHYB2233A
Calcium phosphorylcholine chloride is the main phospholipid component in eukaryotic biofilms. It is also found in commensal or pathogenic bacteria associated with eukaryotes in prokaryotes. This product exhibits a surprising range of immunomodulatory properties and is for research use only.
- Human endogenous metabolite, microbial metabolite
- Component of drug delivery micelles, enhancing cellular internalization and increasing cytotoxicity of cytotoxins in HeLa cells
- Activates nicotinic and muscarinic cholinergic neurotransmission, stimulates α2-adrenergic receptors, and increases plasma glucagon and choline concentrations
- Induces anti-PC and anti-oxLDL antibodies, increases mature B cells in the spleen, and reduces atherosclerotic lesions in mouse models