Learn More
Medchemexpress LLC Calcium phosphorylcholine chloride | 4826-71-5 | 99.0% | 1 KG
SDP
Catalog No. 5000409662
Encompass_Preferred
Calcium phosphorylcholine chloride is a main phospholipid component found in eukaryotic biofilms, existing in commensal or pathogenic bacteria associated with eukaryotes in prokaryotes. It exhibits a surprising range of immunomodulatory properties, making it suitable for various research applications.
  • Main phospholipid component in eukaryotic biofilms.
  • Exhibits immunomodulatory properties.
  • Enhances cellular internalization efficiency as a component of drug delivery micelles.
  • Increases cytotoxicity of cytotoxins in HeLa cells when carried in micelles.
  • Activates nicotinic and muscarinic cholinergic neurotransmission.
  • Stimulates α2-adrenergic receptors.
  • Increases plasma glucagon and choline concentrations.
  • Reduces atherosclerotic lesions in mouse models.
Catalog No. 50-004-09662 Supplier Medchemexpress LLC Supplier No. HYB2233A1KG
Calcium phosphorylcholine chloride is a main phospholipid component found in eukaryotic biofilms, existing in commensal or pathogenic bacteria associated with eukaryotes in prokaryotes. It exhibits a surprising range of immunomodulatory properties, making it suitable for various research applications.
  • Main phospholipid component in eukaryotic biofilms.
  • Exhibits immunomodulatory properties.
  • Enhances cellular internalization efficiency as a component of drug delivery micelles.
  • Increases cytotoxicity of cytotoxins in HeLa cells when carried in micelles.
  • Activates nicotinic and muscarinic cholinergic neurotransmission.
  • Stimulates α2-adrenergic receptors.
  • Increases plasma glucagon and choline concentrations.
  • Reduces atherosclerotic lesions in mouse models.