Learn More
Medchemexpress LLC Calcium phosphorylcholine chloride | 4826-71-5 | 99.0% | 500 G
SDP
Catalog No. 5000410983
Encompass_Preferred
Calcium phosphorylcholine chloride is the primary phospholipid component found in eukaryotic biofilms. It is present in both commensal and pathogenic bacteria that are associated with eukaryotes in prokaryotes, and it demonstrates a notable array of immunomodulatory properties.
  • Serves as a component in drug delivery micelles, enhancing the efficiency of cellular internalization and increasing the cytotoxicity of cytotoxins carried within these micelles in HeLa cells.
  • Activates nicotinic and muscarinic cholinergic neurotransmission, stimulates α2-adrenergic receptors, and increases plasma glucagon and choline concentrations, thereby stimulating peripheral autonomic nervous system activity.
  • Induces the production of anti-PC and anti-oxLDL antibodies, increases mature B cells in the spleen, and reduces atherosclerotic lesions in the aortic root in mouse models.
Catalog No. 50-004-10983 Supplier Medchemexpress LLC Supplier No. HYB2233A500G
Calcium phosphorylcholine chloride is the primary phospholipid component found in eukaryotic biofilms. It is present in both commensal and pathogenic bacteria that are associated with eukaryotes in prokaryotes, and it demonstrates a notable array of immunomodulatory properties.
  • Serves as a component in drug delivery micelles, enhancing the efficiency of cellular internalization and increasing the cytotoxicity of cytotoxins carried within these micelles in HeLa cells.
  • Activates nicotinic and muscarinic cholinergic neurotransmission, stimulates α2-adrenergic receptors, and increases plasma glucagon and choline concentrations, thereby stimulating peripheral autonomic nervous system activity.
  • Induces the production of anti-PC and anti-oxLDL antibodies, increases mature B cells in the spleen, and reduces atherosclerotic lesions in the aortic root in mouse models.