Learn More
Medchemexpress LLC Calcium Phytate | 3615-82-5 | 250 MG
SDP
Catalog No. 5000420650
Encompass_Preferred
Calcium phytate is an insoluble phosphorus-containing compound commonly found in plant-based foods such as cereals and legumes. It can be present in feces and has shown properties to alleviate lead poisoning.
  • Found extensively in plant-based foods such as cereals and legumes
  • Can be present in feces
  • Alleviates lead poisoning
  • Reduces weight loss in lead poisoning models
  • Reduces lead accumulation in the liver and kidneys
  • Reverses toxic effects of lead on liver and kidney weights
Catalog No. 50-004-20650 Supplier Medchemexpress LLC Supplier No. 250MGHYN2582
Calcium phytate is an insoluble phosphorus-containing compound commonly found in plant-based foods such as cereals and legumes. It can be present in feces and has shown properties to alleviate lead poisoning.
  • Found extensively in plant-based foods such as cereals and legumes
  • Can be present in feces
  • Alleviates lead poisoning
  • Reduces weight loss in lead poisoning models
  • Reduces lead accumulation in the liver and kidneys
  • Reverses toxic effects of lead on liver and kidney weights