Learn More
Medchemexpress LLC HY-107929 500mg Medchemexpress, Calcium polystyrene sulfonate CAS:37286-92-3 Purity:>98%
SDP
Catalog No. 501968482
Encompass_Preferred
Medchemexpress, HY-107929 500mg Calcium polystyrene sulfonate CAS:37286-92-3 Calcium polystyrene sulfonate is an ion-exchange resin used for reducing blood levels of potassium. Calcium polystyrene sulfonate is used to treat hyperkalemia in patients with chronic kidney disease (CKD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8482 Supplier Medchemexpress LLC Supplier No. HY107929500MG
Medchemexpress, HY-107929 500mg Calcium polystyrene sulfonate CAS:37286-92-3 Calcium polystyrene sulfonate is an ion-exchange resin used for reducing blood levels of potassium. Calcium polystyrene sulfonate is used to treat hyperkalemia in patients with chronic kidney disease (CKD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.