Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Calcium propionate

Catalog No. 21230250GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-230-250GF 250 g
21-230-1KGF 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-230-250GF Supplier Sigma Aldrich Supplier No. 21230250GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Calcium propionate is generally used as a food preservative in bread
  • It has also been reported to reduce milk fever in dairy cows.calcium propionate was used as an antifungal agent in bread
  • It was also used to investigate the influence of calcium salts on growth, polygalacturonase activity and infection of peach fruit by monilinia fructicola

Specifications

CAS 4075-81-4
Quantity 250 g
Molecular Formula C6H10CaO4
Linear Formula (CH3CH2COO)2 Ca
MDL Number MFCD00036137
Synonym Propionic acid calcium salt
Molecular Weight (g/mol) 186.22
Percent Purity ≥97.0% (dry substance, NT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.