Learn More
Medchemexpress LLC Calcium pyrophosphate | 7790-76-3 | MFCD00015983 | 98.0% | 254.10 g/mol | Ca2P2O7 | 5 G
SDP
Catalog No. 5000299997
Encompass_Preferred
Calcium pyrophosphate is an inorganic biochemical reagent (CAS 7790-76-3) supplied for research use. It is a white, solid powder commonly used to generate calcium pyrophosphate crystals for cellular assays, biochemical studies, and mineralization research. The product is provided with safety and specification documentation to support safe handling and experimental reproducibility.
  • White solid powder suitable for laboratory use.
  • Purity 98.0% as specified by the supplier.
  • Available in 5 g, 10 g, and 50 g pack sizes.
  • Supplied with an SDS and product datasheet for handling and safety information.
  • Used to prepare calcium pyrophosphate crystals for cellular and biochemical assays.
Catalog No. 50-002-99997 Supplier Medchemexpress LLC Supplier No. HYW0199725G
Calcium pyrophosphate is an inorganic biochemical reagent (CAS 7790-76-3) supplied for research use. It is a white, solid powder commonly used to generate calcium pyrophosphate crystals for cellular assays, biochemical studies, and mineralization research. The product is provided with safety and specification documentation to support safe handling and experimental reproducibility.
  • White solid powder suitable for laboratory use.
  • Purity 98.0% as specified by the supplier.
  • Available in 5 g, 10 g, and 50 g pack sizes.
  • Supplied with an SDS and product datasheet for handling and safety information.
  • Used to prepare calcium pyrophosphate crystals for cellular and biochemical assays.