Learn More
Molecular Devices Molecular Devices Membrane Potential Assay Kit, Explorer, Blue Dye, 96/384-Well
Catalog No. 5000014401
Encompass_Preferred
The FLIPR Membrane Potential Assay Kit (Explorer format, Blue) is a homogeneous fluorescence reagent kit for detecting changes in cellular membrane potential in high-throughput screening. The kit simplifies assay setup with prealiquoted dye vials and assay buffer, and is compatible with common plate readers and FLIPR systems.
  • Homogeneous, no-wash fluorescence assay for membrane potential detection.
  • Includes 10 reagent vials and 1 bottle of assay buffer.
  • Blue membrane potential dye formulated for sensitive detection.
  • Suitable for 96-well and 384-well plate formats.
  • Each vial supplies reagent for one plate to simplify assay setup.
  • Intended for research use only; not for diagnostic procedures.
Catalog No. 50-000-14401 Supplier Molecular Devices Supplier No. R8042
Add to Cart
Add to Cart
The FLIPR Membrane Potential Assay Kit (Explorer format, Blue) is a homogeneous fluorescence reagent kit for detecting changes in cellular membrane potential in high-throughput screening. The kit simplifies assay setup with prealiquoted dye vials and assay buffer, and is compatible with common plate readers and FLIPR systems.
  • Homogeneous, no-wash fluorescence assay for membrane potential detection.
  • Includes 10 reagent vials and 1 bottle of assay buffer.
  • Blue membrane potential dye formulated for sensitive detection.
  • Suitable for 96-well and 384-well plate formats.
  • Each vial supplies reagent for one plate to simplify assay setup.
  • Intended for research use only; not for diagnostic procedures.