Learn More
Scytek Laboratories Calcium Stain Kit (Modified Von Kossa): Large
SDP
Catalog No. NC1969831
Encompass_Preferred
The Calcium Stain Kit (Modified Von Kossa) is intended for use in the histological visualization of calcium deposits in paraffin sections.Calcium in mass deposits: BlackCalcium in dispersed deposits: GrayNuclei: RedCytoplasm: Light PinkKit contents: SNV125, Silver Nitrate Solution (5%), 125 ml.STB125, Sodium Thiosulfate Solution (5%), 125 ml.NFS125, Nuclear Fast Red Solution, 125 ml.
This item is not returnable. View return policy
Catalog No. NC1969831 Supplier Scytek Laboratories Supplier No. CVK1
The Calcium Stain Kit (Modified Von Kossa) is intended for use in the histological visualization of calcium deposits in paraffin sections.Calcium in mass deposits: BlackCalcium in dispersed deposits: GrayNuclei: RedCytoplasm: Light PinkKit contents: SNV125, Silver Nitrate Solution (5%), 125 ml.STB125, Sodium Thiosulfate Solution (5%), 125 ml.NFS125, Nuclear Fast Red Solution, 125 ml.