Learn More
Scytek Laboratories Calcium Stain Kit (Modified Von Kossa): Small
SDP
Catalog No. NC1969834
Encompass_Preferred
The Calcium Stain Kit (Modified Von Kossa) is intended for use in the histological visualization of calcium deposits in paraffin sections.Calcium in mass deposits: BlackCalcium in dispersed deposits: GrayNuclei: RedCytoplasm: Light PinkKit contents: SNV030, Silver Nitrate Solution (5%), 30 ml.STB030, Sodium Thiosulfate Solution (5%), 30 ml.NFS030, Nuclear Fast Red Solution, 30 ml.
This item is not returnable. View return policy
Catalog No. NC1969834 Supplier Scytek Laboratories Supplier No. CVK2
The Calcium Stain Kit (Modified Von Kossa) is intended for use in the histological visualization of calcium deposits in paraffin sections.Calcium in mass deposits: BlackCalcium in dispersed deposits: GrayNuclei: RedCytoplasm: Light PinkKit contents: SNV030, Silver Nitrate Solution (5%), 30 ml.STB030, Sodium Thiosulfate Solution (5%), 30 ml.NFS030, Nuclear Fast Red Solution, 30 ml.