Learn More
SPEX CERTIPREP LLC 3P Calcium at 10,000 ug/mL in 4% HNO3, 100 mL
Catalog No. NC0911079
Encompass_Preferred
(HPS by Zeptometrix) Calcium (Ca) ICP Single-Component Standard at 10,000 ug/mL in 4% HNO3.
Catalog No. NC0911079 Supplier SPEX CERTIPREP LLC 3P Supplier No. 10M91100
(HPS by Zeptometrix) Calcium (Ca) ICP Single-Component Standard at 10,000 ug/mL in 4% HNO3.