Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium standard for AAS, 1.000 μg/L Ca+2 in hydrochloric acid, Traceable to BAM, MilliporeSigma™ Supelco™

Catalog No. 111020647 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0647 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0647 Supplier MilliporeSigma Supelco Supplier No. 21049100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 10043-52-4
Linear Formula CaCl2
MDL Number MFCD00010903
Synonym Calcium chloride solution
Type Analytical Standard
Quantity 100 mL
Formula Weight 110.98
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 1.000 g/L Ca +2 in. hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.