Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Standard, Ultra Scientific

Catalog No. USICP020
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-ICP-020 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-ICP-020 Supplier Agilent Technologies Supplier No. ICP020
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10043-52-4 , 7697-37-2 , 7732-18-5
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Packaging Bottle
CAS Max % 94.0, 5.0, 1.0
Analyte or Component Names Calcium,water,nitric acid
Element Nitric Acid, Ca
Solvent or Matrix Water/Dilute Nitric Acid
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Calcium Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.