Learn More
Hach Company Calcium Standard Solution, 10 mg/L, 100 mL
Supplier:  Hach Company 2305442
Encompass
ISE Standard Solution. Calcium, 10.00 mg/L ±0.14 mg/L as Ca2+ (NIST). 100 mL bottle
Catalog No. 50-199-5758