Learn More
Hach Company Calcium Standard Solution, 100 mg/L, 500 mL
Catalog No. 501997040 Shop All Hach Company Products
Encompass
ISE Standard Solution. Calcium, 100 mg/L ±1 mg/L as Ca2+ (NIST). 500 mL bottle
Catalog No. 50-199-7040 Supplier Hach Company Supplier No. 4457649
ISE Standard Solution. Calcium, 100 mg/L ±1 mg/L as Ca2+ (NIST). 500 mL bottle