Learn More
Medchemexpress LLC Calcium stearate | 1592-23-0 | MFCD00036390 | 50 G
SDP
Catalog No. 5000410995
Encompass_Preferred
Calcium stearate is a long-chain dietary saturated fatty acid found in many animal and vegetable fats and oils. It is classified as an endogenous metabolite with a molecular weight of 607.02 and a formula of C36H70CaO4. This product appears as a white to off-white solid.
  • Suitable for research applications.
  • Insoluble in ethanol.
  • Ships at room temperature in continental US.
  • Store at room temperature in a dry, cool environment.
  • Can be stored in solvent at -80°C for 1 year or -20°C for 6 months.
Catalog No. 50-004-10995 Supplier Medchemexpress LLC Supplier No. HYB2219A50G
Calcium stearate is a long-chain dietary saturated fatty acid found in many animal and vegetable fats and oils. It is classified as an endogenous metabolite with a molecular weight of 607.02 and a formula of C36H70CaO4. This product appears as a white to off-white solid.
  • Suitable for research applications.
  • Insoluble in ethanol.
  • Ships at room temperature in continental US.
  • Store at room temperature in a dry, cool environment.
  • Can be stored in solvent at -80°C for 1 year or -20°C for 6 months.