Learn More
Medchemexpress LLC Calcium sulfate, anhydrous | 7778-18-9 | MFCD00010912 | 99.4% | 136.14 g/mol | CaSO4 | 100 G
SDP
Catalog No. 5000301853
Encompass_Preferred
Anhydrous calcium sulfate (CAS 7778-18-9) is a white, inorganic powder used as a reagent, desiccant, and component in material formulations where a water-free sulfate is required. The material offers stable handling properties and is intended for laboratory and formulation use when low moisture content is critical.
  • High anhydrous purity suitable for reagent-grade applications.
  • Low moisture content when stored sealed.
  • Stable solid form for easy handling and accurate weighing.
  • Effective desiccant for drying solvents and samples.
  • Compatible with material science and biomedical formulation workflows.
  • Available in laboratory-scale packaging for research use.
Catalog No. 50-003-01853 Supplier Medchemexpress LLC Supplier No. HYW019901B100G
Anhydrous calcium sulfate (CAS 7778-18-9) is a white, inorganic powder used as a reagent, desiccant, and component in material formulations where a water-free sulfate is required. The material offers stable handling properties and is intended for laboratory and formulation use when low moisture content is critical.
  • High anhydrous purity suitable for reagent-grade applications.
  • Low moisture content when stored sealed.
  • Stable solid form for easy handling and accurate weighing.
  • Effective desiccant for drying solvents and samples.
  • Compatible with material science and biomedical formulation workflows.
  • Available in laboratory-scale packaging for research use.