Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium Sulfate, Dihydrate
SCI_ED

Catalog No. S25230A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade - Powder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25230A 500 g Reagent Grade - Powder
S25230 100 g Reagent Grade - Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25230A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10101-41-4 , 7778-16-9

Specifications

Color White
Quantity 500 g
Assay Percent Range >95% w/w
Molecular Formula CaH4O6S
Linear Formula CaSO4·2H2O
MDL Number MFCD00149625
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O
IUPAC Name calcium dihydrate sulfate
Molecular Weight (g/mol) 172.16
PubChem CID 24928
ChEBI CHEBI:32583
Formula Weight 172.17
Grade Reagent
Packaging Reagent Grade - Powder
Density 2.32g/mL
Chemical Name or Material Calcium Sulfate, Dihydrate
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.