Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Sulfate Hemihydrate, Solstice

Catalog No. 6001925 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-25 1 kg
60-019-26 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-019-25 Supplier Solstice Supplier No. 120901KG
Add to Cart
Add to Cart

Purum,≥97.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-76-1
Molecular Formula Ca2H2O9S2
Molecular Weight (g/mol) 290.283
MDL Number MFCD00799067
InChI Key ZOMBKNNSYQHRCA-UHFFFAOYSA-J
Synonym calcium sulfate hemihydrate, gypsum hemihydrate, calcium sulphate hemihydrate, unii-3rw091j48v, crystacal, densite, hemihydrate gypsum, densite gypsum, gipshalbhydrat, p 23 sulfate
PubChem CID 3033839
ChEBI CHEBI:32584
IUPAC Name dicalcium;disulfate;hydrate
SMILES O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ca+2].[Ca+2]

Specifications

Quantity 1 kg
Linear Formula CaSO4 · 0.5H2O
UN Number NONH for all modes of transport
Formula Weight 145.15g/mol
Percent Purity ≥97%
Grade purum
Chemical Name or Material Calcium Sulfate Hemihydrate
EINECSNumber 231-900-3
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.