Learn More
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 25 kg |
| Linear Formula | CaSO4 · 0.5H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 145.15g/mol |
| Percent Purity | ≥97% |
| Grade | purum |
| Chemical Name or Material | Calcium Sulfate Hemihydrate |
Safety and Handling
| EINECSNumber | 231-900-3 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.