Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Thioglycolate Trihydrate 94.0+%, TCI America™
SDP

Catalog No. T090225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T090225G 25 g
T0902500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T090225G Supplier TCI America Supplier No. T090225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5793-98-6
Molecular Formula C4H14CaO7S2
Molecular Weight (g/mol) 278.347
MDL Number MFCD00043437
InChI Key YKVKKJPPSKLAFH-UHFFFAOYSA-N
Synonym calciumthioglycolatetrihydrate
PubChem CID 131881455
IUPAC Name calcium;2-sulfanylacetic acid;trihydrate
SMILES C(C(=O)O)S.C(C(=O)O)S.O.O.O.[Ca]

Specifications

Color White
Quantity 25 g
Formula Weight 130.17
Percent Purity ≥94.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium Thioglycolate Trihydrate
EINECSNumber (2)-1363
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.