Learn More
Strem, An Ascensus Company CAS# 55120-75-7. 10g. Calcium trifluoromethanesulfonate, min. 96% (Calcium triflate). MFCD00143911
SDP
Catalog No. 5090110695
Encompass
CAS# 55120-75-7. 10g. Calcium trifluoromethanesulfonate, min. 96% (Calcium triflate). MFCD00143911. Molecular Weight: 338.21. Molecular Formula: Ca(CF3SO3)2. Color/form: white pwdr. Strem# 20-5000. http://www.strem.com/catalog/v/20-5000/
Catalog No. 50-901-10695 Supplier Strem, An Ascensus Company Supplier No. 20500010G
CAS# 55120-75-7. 10g. Calcium trifluoromethanesulfonate, min. 96% (Calcium triflate). MFCD00143911. Molecular Weight: 338.21. Molecular Formula: Ca(CF3SO3)2. Color/form: white pwdr. Strem# 20-5000. http://www.strem.com/catalog/v/20-5000/