Learn More
Strem, An Ascensus Company CAS# 7790-75-2. 100g. Calcium tungstate, 98%. MFCD00010913
SDP
Catalog No. 5090110687
Encompass
CAS# 7790-75-2. 100g. Calcium tungstate, 98%. MFCD00010913. Molecular Weight: 287.93. Molecular Formula: CaWO4. Color/form: white to buff pwdr. Strem# 20-2052. http://www.strem.com/catalog/v/20-2052/
Catalog No. 50-901-10687 Supplier Strem, An Ascensus Company Supplier No. 202052100G
CAS# 7790-75-2. 100g. Calcium tungstate, 98%. MFCD00010913. Molecular Weight: 287.93. Molecular Formula: CaWO4. Color/form: white to buff pwdr. Strem# 20-2052. http://www.strem.com/catalog/v/20-2052/