Learn More
MSC ZEP All-Purpose Cleaner, Liquid, Acidic, Calcium Lime Remover, 1 gal
Catalog No. 502867547
Encompass
This all-purpose cleaner is a liquid degreaser with an acidic scent, formulated to efficiently eliminate lime scale and haze from kitchen equipment and utensils. It is safe for use on stainless steel, aluminum, glass, plastic, and ceramic surfaces.
  • Efficiently eliminates lime scale and haze
  • Safe for use on stainless steel, aluminum, glass, plastic, and ceramic surfaces
  • Contains orthophosphoric acid (30-50%) and ethoxylated alcohols (1-3%)
  • Features a low pH of 1-1.5
  • Water soluble with a density of 1.259 g/cm³
Catalog No. 50-286-7547 Supplier MSC Supplier No. 17643552
This all-purpose cleaner is a liquid degreaser with an acidic scent, formulated to efficiently eliminate lime scale and haze from kitchen equipment and utensils. It is safe for use on stainless steel, aluminum, glass, plastic, and ceramic surfaces.
  • Efficiently eliminates lime scale and haze
  • Safe for use on stainless steel, aluminum, glass, plastic, and ceramic surfaces
  • Contains orthophosphoric acid (30-50%) and ethoxylated alcohols (1-3%)
  • Features a low pH of 1-1.5
  • Water soluble with a density of 1.259 g/cm³