Learn More
Calcofluor White Stain, 100 mL, G-Biosciences
SCI_ED
Catalog No. S001147
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
S001147 100 mL
1 options
Catalog No. S001147 Supplier G-Biosciences Supplier No. 7862246
Add to Cart
Add to Cart
Calcofluor White Stain is used for the detection of many yeasts and pathogenic fungi such as Microsporidium, Acanthamoeba, Pneumocystis, Naegleria, and Balamuthia species.
  • Calcofluor White Stain binds with cellulose and chitin components of the cell walls of fungi and other organisms.
Specifications
Quantity 100 mL
Product Type Calcofluor White Stain