Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcofluor White Stain, 100 mL
Catalog No. NC1021099
Encompass
A fluorescent stain for rapid detection of yeasts, fungi and parasitic organisms. Calcofluor White is a non-specific fluorochrom that binds to cellucose and chitin in cell walls.
Catalog No. NC1021099 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 18909100MLF
A fluorescent stain for rapid detection of yeasts, fungi and parasitic organisms. Calcofluor White is a non-specific fluorochrom that binds to cellucose and chitin in cell walls.