Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcofluor White Stain, Sigma-Aldrich

Catalog No. 01258921 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-921 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-921 Supplier MilliporeSigma Supelco Supplier No. 18909100MLF
Add to Cart
Add to Cart

Calcofluor White Stain is a stilbene compound that has additional triazinyl and phenyl aromatic groups. It is also known as cellufluor.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcofluor White Stain
Color Black/Blue to Blue
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.