Learn More
Medchemexpress LLC Calconcarboxylic acid | 3737-95-9 | MFCD00004078 | 438.41 | C21H14N2O7S | 10g
SDP
Catalog No. 5000269681
Encompass_Preferred
Calconcarboxylic acid an azo dye acts as a silver-ion sensitizer to stain protein in SDS-PAGE gels Calconcarboxylic acid increases silver binding on protein bands or spots by the formation of a silver-dye complex and also increases the reducing power o
Catalog No. 50-002-69681 Supplier Medchemexpress LLC Supplier No. HYY070010G
Calconcarboxylic acid an azo dye acts as a silver-ion sensitizer to stain protein in SDS-PAGE gels Calconcarboxylic acid increases silver binding on protein bands or spots by the formation of a silver-dye complex and also increases the reducing power o