Learn More
Aviva Systems Biology Corp CALCR Antibody - C-terminal region (ARP59632_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59632_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CALCR. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CALCR. Alternate names: CRT, CTR, CTR1, CT-RApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human
Catalog No. 50-286-764