Learn More
Vernier™ Calculator Link Cable
SDPSCI_ED
Catalog No. S16264ND Shop All Vernier Software Products
Change view
Click to view available options
Length (English):
16 in.
Catalog No. Length (English)
S16264ND 16 in.
1 options
Catalog No. S16264ND Supplier Vernier™ Supplier No. TICLC
Includes
Calculator Link Cable
This 16 in. cable is used to connect a LabPro or CBL 2 to the I/O port of a TI-84 Plus calculator. The cable has the small I/O plug on both ends. The cable can also be used to connect a graphing calculator to the I/O port on a Go!Motion, CBR 2, or another calculator. This cable is included with a CBL 2 or LabPro interface.
  • The cable has the small I/O plug on both ends.
  • The cable can also be used to connect a graphing calculator to the I/O port on a Go!Motion, CBR 2, or another calculator.
  • This cable is included with a CBL 2 or LabPro interface.
Warranty and Services
5-year limited warranty
Specifications
For Use With (Equipment) CBL 2 and LabPro
Length (English) 16 in.
Length (Metric) 40.64 cm
For Use With (Application) Educational use only