Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcum Gluconate Monhydra500MG
Catalog No. 502898418
Encompass
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Catalog No. 50-289-8418 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PHR2670500MG
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.